Tetrallyltin ≥97.0% (by GC)

Leverancier: TCI EUROPE
Gevaar

TCIAT2009-5GEA 142 EUR
TCIAT2009-5G TCIAT2009-1G
Tetrallyltin ≥97.0% (by GC)
Tetrallyltin
Formule: C₁₂H₂₀Sn
MW: 283 g/mol
Kookpunt: 70°C
Vlampunt: 75°C
MDL-nummer: MFCD00008637
CAS-nummer: 7393-43-3
EINECS: 230-987-5
UN: 2788
ADR: 6.1,III
https://be.vwr.com/stibo/web/std.lang.all/59/50/28995950.jpg

Bestel nu

Specificaties testresultaten

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Gravimetric Method) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR