(Pentafluorophenyl)acetic acid ≥98.0% (by GC, titration analysis)

Leverancier: TCI EUROPE
Waarschuwing

TCIAP1004-1GEA 48.4 EUR
TCIAP1004-1G
(Pentafluorophenyl)acetic acid ≥98.0% (by GC, titration analysis)
(Pentafluorophenyl)acetic acid
Formule: C₈H₃F₅O₂
MW: 226,1 g/mol
Smeltpunt: 140,5…141.5 °C
MDL-nummer: MFCD00004316
CAS-nummer: 653-21-4
EINECS: 211-497-0
https://be.vwr.com/stibo/web/std.lang.all/16/39/28991639.jpg

Bestel nu

Specificaties testresultaten

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 108.0 to 111.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR