Benzenesulphonyl chloride ≥99.0% (by GC, titration analysis) for determination of hippuric acid

Leverancier: TCI EUROPE
Gevaar

TCIAB1211-25GEA 25.3 EUR
TCIAB1211-25G
Benzenesulphonyl chloride ≥99.0% (by GC, titration analysis) for determination of hippuric acid
Benzenesulphonyl chloride
Formule: C₆H₅ClO₂S
MW: 176,62 g/mol
Kookpunt: 252°C
Smeltpunt: 15 °C
Vlampunt: 132°C
MDL-nummer: MFCD00007426
CAS-nummer: 98-09-9
EINECS: 202-636-6
UN: 2225
ADR: 8,III
Merck Index: 13,01072
https://be.vwr.com/stibo/web/std.lang.all/30/47/28933047.jpg

Bestel nu

Specificaties testresultaten

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %
Suitability test to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR